Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Willebrand Factor Antibody (SPM577), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234810MFV610
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow CytometrySpecifications
Von Willebrand Factor | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
SPM577 | |
Flow Cytometry | |
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
Recombinant human Von Willebrand Factor fragment (Uniprot: P04275 ) | |
0.1 mL | |
Cancer | |
7450 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction