Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/1465), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $536.00
Specifications
Antigen | vWF-A2 |
---|---|
Clone | VWF/1465 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25331420
![]() |
Novus Biologicals
NBP25331420UG |
20 μg |
Each for $268.00
|
|
|||||
NB253314100
![]() |
Novus Biologicals
NBP253314100UG |
100 μg |
Each for $536.00
|
|
|||||
NB253314200
![]() |
Novus Biologicals
NBP253314200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
vWF-A2 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
F8VWF, VWD, VWF von Willebrand factor | |
VWF | |
IgG1 | |
Protein A or G purified | |
250 kDa |
VWF/1465 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
7450 | |
Recombinant fragment of human vWF protein (aa1815-1939) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title