Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand Factor Monoclonal Antibody (VW92-3), QED Bioscience™
SDP

Catalog No. 502277899
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7899 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7899 Supplier Leinco Technologies Inc Supplier No. 19818200UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

von Willebrand Factor Monoclonal antibody specifically detects von Willebrand Factor in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen von Willebrand Factor
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VW92-3
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and no preservative; pH 7.4
Gene VWF
Gene Accession No. P04275
Gene Alias 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312
Host Species Mouse
Immunogen Human plasma von Willebrand Factor.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.