Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Willebrand Factor Mouse anti-Human, Clone: rVWF/1465, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30721620UG
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Von Willebrand Factor | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunoprecipitation 0.5-1 ug/500 ug protein lysate, Immunohistochemistry-Paraffin 1-2 ug/ml | |
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
rVWF/1465 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
7450 | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction