Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Mouse anti-Human, Clone: VWF/2480, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Von Willebrand Factor
Clone VWF/2480
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121872
SDP
View Documents
Novus Biologicals
NBP30719920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Von Willebrand Factor
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Recombinant full-length human Von Willebrand Factor protein
Primary
Store at 4°C.
VWF/2480
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
7450
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.