Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand Factor, Rabbit anti-Human, Mouse, Rat, Polyclonal, Millipore Sigma™

Catalog No. AB735625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB735625UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB735625UG Supplier MilliporeSigma Supplier No. AB735625UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-von Willebrand Factor Antibody is validated for use in ELISA, Immunohistochemistry (IHC) for the detection of von Willebrand Factor.

von Willebrand Factor (vWF) is synthesized by endothelial cells, causing adhesion of platelets to injured vessel walls and functions as a carrier and stabilizer for coagulation of Factor VIII. It is one of the most widely used endothelial cell markers for studying angiogenesis / neovascularization. Millipore's von Willebrand Factor antibody reacts specifically with the endothelial cells of blood vessels and is a useful marker for the identification of endothelial lineage in tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen von Willebrand Factor
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA: A 1:1000-1:5000 dilution of a previous lot was used in direct ELISA.Optimal working dilutions must be determined by the end user.
Formulation Purified rabbit immunoglobulin in buffer containing 0.02 M PBS pH 7.6, 0.25 M NaCl with 0.1% sodium azide.
Gene Alias Factor VIII Related Antigen
Gene Symbols VWF;F8VWF;F8;vWF;VWD
Host Species Rabbit
Immunogen Human von Willebrand Factor purified from plasma.
Molecular Weight of Antigen 250 kDa
Purification Method Protein A Purfied
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NM_000552.3
Test Specificity AB7356 is specific for vWF by ELISA against the human protein and by immunohistochemistry. The antibody is reactive in paraffin embedded tissues on human, rat, and mouse. Other species not tested. Antibody has not been evaluated by western blot analysis for reactivity to vWF.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8ºC from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.