Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand factor (vWF) Mouse anti-Human, Clone: AvW3, MilliporeSigma™

Catalog No. MABS215125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS215125 25 μg
MABS2151100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS215125 Supplier MilliporeSigma Supplier No. MABS215125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

von Willebrand factor (vWF) Monoclonal antibody specifically detects von Willebrand factor (vWF) in Human samples. It is validated for ELISA, Inhibits Activity/Function, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen von Willebrand factor (vWF)
Applications ELISA, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone AvW3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without preservatives.
Host Species Mouse
Immunogen Purified human vWF antigen II (aa 23-763).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.