Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vopp1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571488
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571488 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571488 Supplier Invitrogen™ Supplier No. PA571488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vopp1 Polyclonal Antibody for Western Blot

Increases the transcriptional activity of NFKB1 by facilitating its nuclear translocation, DNA-binding and associated apoptotic response, when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vopp1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene VOPP1
Gene Accession No. Q8R1C3, Q96AW1
Gene Alias AW146242; ECOP; EGFR-coamplified and overexpressed protein; GASP; Glioblastoma-amplified secreted protein; hypothetical protein DKFZp564K0822; putative NF-kappa-B-activating protein 055N; RGD1306494; Vesicular, overexpressed in cancer, prosurvival protein 1; VOPP1
Gene Symbols VOPP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 65-95 amino acids from the Central region of human ECOP.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232023, 81552
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.