Learn More
Corning™ LSE™ Digital Vortexer Platform
Corning™ Vortexer Platform for use with Corning™ LSE Multiposition Digital Vortexer.
$279.50 - $484.50
Specifications
For Use With (Application) General Purpose Mixing
For Use With (Equipment) LSE Multiposition Digital Vortexer
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
07-202-206
View Documents Promotion Details
Corning™
48010
Platform
Each for $279.50
Only null left
Add to Cart
07-202-207
View Documents Promotion Details
Corning™
48011
Platform with non-stick mat
Each for $290.50
Only null left
Add to Cart
07-202-208
View Documents Promotion Details
Corning™
48012
Spring Loaded Platform
Each for $484.50
Only null left
Add to Cart
Description
Description
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
  • Five-year warranty
Specifications
Specifications
General Purpose Mixing
LSE Multiposition Digital Vortexer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.