Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Vortexer Mixer, Blue

Catalog No. 14127522
Change view
Click to view available options
Plug Type:
AUS Plug
CE
JP Plug
UK Plug
US Plug
Electrical Requirements:
110/120 V, 50/60 Hz
220/240 V, 50/60 Hz
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Electrical Requirements
14-127-522 UK Plug 220/240 V, 50/60 Hz
14-127-520 US Plug 110/120 V, 50/60 Hz
14-127-521 CE 220/240 V, 50/60 Hz
14-127-524 JP Plug 220/240 V, 50/60 Hz
14-127-523 AUS Plug 220/240 V, 50/60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 14-127-522 Supplier Heathrow Scientific™ Supplier No. HS120214
Only null left
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

The Vortexer is a multi-purpose, and maximum convenience unit. This sleek and modern designed Vortexer Mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.

The innovative design makes maximum use of the space available, providing holding slots for no less than six different sizes of tube and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.
  • Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed for cleaning purposes
  • US Design Patent No. D787,085, US Utility Patent No. 9895670

Specifications

Orbit 3.7 mm
Load Bearing Capacity (Metric) 135 g
Speed Range 1000 to 3000 rpm
Electrical Requirements 220/240 V, 50/60 Hz
Voltage 220/240 V
Wattage 44 W
Amperage 0.4 A
Tube Capacity 50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL
Product Type Vortexing unit
Load Bearing Capacity (English) 4.8 oz.
Certifications/Compliance SGS-US-C, CE, CB Scheme, RoHS 2, PSE, WEEE
Plug Type UK Plug
Frequency 50/60 Hz
Material ABS Base, Nylon Platform
Depth (English) 7.41 in.
Depth (Metric) 19 cm
Dimensions (L x W) 6.7 x 7.5 in.
Dimensions (L x W x H) 6.7 x 7.5 x 7.8 in.
Height (English) 7.68 in.
Height (Metric) 19.7 cm
Length (English) 6.7 in.
Length (English) Platform 5.51 in.
Length (Metric) 17.1 cm
Length (Metric) Platform 140 mm
Width (English) 6.67 in.
Width (English) Platform 4.5 in.
Width (Metric) 17.1 cm
Width (Metric) Platform 11.43 cm
Shipping Weight (English) 11.12 lb.
Shipping Weight (Metric) 5 kg
Weight (English) 8.66 lb.
Weight (Metric) 3.93 kg
Color Blue
Controller Type 3 position slider (top) intermittent, (middle) off, (bottom) continuous
Description Vortexer Mixer 220/240 V, UK Plug
Model HS120214
For Use With (Equipment) Tubes, Assay Plates
Includes 1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads
Motor Type Brushless
Operating Modes 3-Modes: 1000, 2000, and 3000 rpm
Warranty and Service Offering 5 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.