Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VP2 Mouse anti-Human, Clone:4E5A4, Abnova™
Mouse monoclonal antibody raised against recombinant human VP2.
Supplier: Abnova Corporation MAB17262
Specifications
VP2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
VP2 | |
Recombinant protein corresponding to amino acid 296-438 of human VP2 from E. coli. | |
11293627 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
4E5A4 | |
major capsid protein | |
In PBS (0.05% sodium azide) | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction