Learn More
Description
Specifications
Specifications
| Antigen | VP26B |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | MGC10485, Pep8b, vacuolar protein sorting 26 homolog B (S. pombe), vacuolar protein sorting 26 homolog B (yeast), vacuolar protein sorting-associated protein 26B, Vesicle protein sorting 26B |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human VP26B (NP_443107.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
