Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VP26B Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095430 20 μL
NB095429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095430 Supplier Novus Biologicals Supplier No. NBP30329420UL

Rabbit Polyclonal Antibody

VP26B Polyclonal specifically detects VP26B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VP26B
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias MGC10485, Pep8b, vacuolar protein sorting 26 homolog B (S. pombe), vacuolar protein sorting 26 homolog B (yeast), vacuolar protein sorting-associated protein 26B, Vesicle protein sorting 26B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human VP26B (NP_443107.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112936
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.