Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VP3 Goose parvovirus Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986059
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6059 Supplier Bioss Supplier No. BS4499R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The GPV VP3-encoding gene is 1605 bp in length, and it encodes a 534 amino acid protein with a predicted molecular mass of 59.9 kDa. The VP3 protein of goose parvovirus (GPV) or Muscovy duck parvovirus (MDPV), a major structural protein, can induce neutralizing antibodies in geese and ducks, but monoclonal antibodies (MAbs) against VP3 protein has never been characterized.

Specifications

Antigen VP3 Goose parvovirus
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Goose parvovirus VP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.