Learn More
VPAC2 Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: VPAC2 tansfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | VPAC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Vipr2 |
| Gene Accession No. | P41587 |
| Gene Alias | C16DUPq36.3; DUP7q36.3; helodermin-preferring VIP receptor; PACAP type III receptor; Pacap type-3; PACAP3; PACAP-R3; PACAP-R-3; PACAPR-3; pituitary adenylate cyclase-activating polypeptide type III receptor; Pvr3; SCZD16; tcag7.970; vasoactive intestinal peptide receptor 2; vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP receptor subtype 2; Vip type-2 receptor; Vip2; Vip2 receptor; VIP2R; Vipr2; Vip-r2; VIP-R-2; VPAC2; VPAC2 receptor; VPAC2R; VPCAP2R |
| Gene Symbols | Vipr2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.