Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPAC2R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980134
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0134 Supplier Bioss Supplier No. BS0197R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Vasoactive Intestinal Polypeptide Receptor 2 (VPAC2) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.

Specifications

Antigen VPAC2R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Vipr2
Gene Accession No. P41587
Gene Alias C16DUPq36.3; DUP7q36.3; Helodermin-preferring VIP receptor; PACAP type III receptor; Pacap type-3; PACAP3; PACAP-R3; PACAP-R-3; PACAPR-3; Pituitary adenylate cyclase-activating polypeptide type III receptor; Pvr3; SCZD16; tcag7.970; vasoactive intestinal peptide receptor 2; Vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP receptor subtype 2; Vip type-2 receptor; Vip2; Vip2 receptor; VIP2R; Vipr2; Vip-r2; VIP-R-2; VPAC2; VPAC2 receptor; VPAC2R; VPCAP2R
Gene Symbols Vipr2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from rat VIP receptor II.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7434
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.