Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPREB1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299054PCP
Description
VPREB1 Polyclonal antibody specifically detects VPREB1 in Mouse samples. It is validated for Western BlotSpecifications
VPREB1 | |
Polyclonal | |
Western Blot | |
CD179 antigen-like family member A, CD179a, IGI, immunoglobulin iota chain, pre-B lymphocyte 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human VPREB1 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
7441 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction