Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPREB3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP147213
Description
VPREB3 Polyclonal specifically detects VPREB3 in Human, Mouse, Rat, Porcine samples. It is validated for Peptide ELISA.Specifications
| VPREB3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody recognizes and epitope within the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:16000 | |
| NP_037510.1 | |
| VPREB3 | |
| Peptide with sequence EAHNACVLTISPVQ corresponding to internal region according to NP_037510.1. | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| 29802 | |
| Human, Mouse, Rat, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction