Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS11 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204092IR
Description
VPS11 Polyclonal antibody specifically detects VPS11 in Human samples. It is validated for Western BlotSpecifications
VPS11 | |
Polyclonal | |
Western Blot | |
hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human Vacuolar Protein Sorting 11 using the numbering given in entry NP_068375.3 (GeneID 55823). | |
0.1 mL | |
Autophagy, Zinc Finger | |
55823 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction