Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558629 Supplier Invitrogen™ Supplier No. PA558629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQHCFESYSE SDADCPTCLP ENRKVMDMIR AQEQKRDLHD QFQHQLKCSN DSFSVIADYF GRGVFNKLTL LTDPPTARLT SSLEAGLQRD LLMHSR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS11
Gene Accession No. Q9H270
Gene Alias 1200011A11Rik; END1; HLD12; hVPS11; PEP5; PP3476; RING finger protein 108; RNF108; vacuolar protein sorting 11 (yeast); vacuolar protein sorting 11 homolog; vacuolar protein sorting 11 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 11 homolog; VPS11; VPS11 CORVET/HOPS core subunit; VPS11, CORVET/HOPS core subunit
Gene Symbols VPS11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS11. Recombinant protein control fragment (Product #RP-97405).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.