Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS11 Rabbit anti-Human, Mouse, Clone: 8L3D8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | VPS11 |
|---|---|
| Clone | 8L3D8 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
VPS11 Monoclonal antibody specifically detects VPS11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VPS11 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VPS11 (Q9H270). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8L3D8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Zinc Finger | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 55823 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title