Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS13B Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282052V
This item is not returnable.
View return policy
Description
VPS13B Polyclonal antibody specifically detects VPS13B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| VPS13B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 405 | |
| cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast) | |
| VPS13B antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human VPS13B. The immunogen is located within amino acids 90 - 140 of VPS13B. Amino Acid Squence: TNRSTAESTKSSIKPRR | |
| 0.1 mL | |
| Signal Transduction | |
| 157680 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction