Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS13B Antibody (OTI6F6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274853IR
Description
VPS13B Monoclonal specifically detects VPS13B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VPS13B | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VPS13B | |
Recombinant protein fragment corresponding to amino acids 91-346 of human VPS13B (NP_858047) produced in E.coli. | |
0.1 mL | |
157680.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6F6 | |
Western Blot | |
cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction