Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS13B Antibody (OTI6F6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246350
Description
Ensure accurate, reproducible results in Western Blot
VPS13B Monoclonal specifically detects VPS13B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VPS13B | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast) | |
Mouse | |
47 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI6F6 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
VPS13B | |
Human recombinant protein fragment corresponding to amino acids 91-346 of human VPS13B(NP_858047) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
157680 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction