Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS13B Mouse anti-Human, Clone: OTI6D1, lyophilized, TrueMAB™
SDP

Catalog No. 501668492
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8492 Supplier OriGene Supplier No. CF807803
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.

Specifications

Antigen VPS13B
Applications Western Blot
Classification Monoclonal
Clone OTI6D1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene VPS13B
Gene Accession No. Q7Z7G8
Gene Alias CHS1, COH1
Gene Symbols VPS13B
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 91-346 of human VPS13B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157680
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.