Learn More
VPS13C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | VPS13C |
|---|---|
| Concentration | 333 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8756
![]()
|
Proteintech Group Inc
286761AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The VPS13C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS13C. This antibody recognizes human, mouse antigen. The VPS13C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| VPS13C | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 333 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 54832 | |
| Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.