Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS13D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562444 Supplier Invitrogen™ Supplier No. PA562444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WSGGFEVNKN NSFHINMRDT LGKCFFLRVE ITLRGATYRI SFSDTDQLPP PFRIDNFSKV PVVFTQHGVA EPRLRTEVKP M Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Functions in promoting mitochondrial clearance by mitochondrial autophagy (mitophagy), also possibly by positively regulating mitochondrial fission. Mitophagy plays an important role in regulating cell health and mitochondrial size and homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS13D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS13D
Gene Accession No. Q5THJ4
Gene Alias FLJ10619; KIAA0453; vacuolar protein sorting 13 D (yeast); vacuolar protein sorting 13 homolog D; vacuolar protein sorting 13 homolog D (S. cerevisiae); vacuolar protein sorting 13D; vacuolar protein sorting-associated protein 13D; VPS13D
Gene Symbols VPS13D
Host Species Rabbit
Immunogen Recombinant protein fragment of Human VPS13D (Product #RP-93989)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.