Learn More
Invitrogen™ VPS13D Polyclonal Antibody

Description
Immunogen sequence: WSGGFEVNKNN SFHINMRDTL GKCFFLRVEI TLRGATYRIS FSDTDQLPPP FRIDNFSKVP VVFTQHGVAE PRLRTEVKPM Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
Specifications
Specifications
| Antigen | VPS13D |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | VPS13D |
| Gene Accession No. | Q5THJ4 |
| Gene Alias | FLJ10619; KIAA0453; vacuolar protein sorting 13 D (yeast); vacuolar protein sorting 13 homolog D; vacuolar protein sorting 13 homolog D (S. cerevisiae); vacuolar protein sorting 13D; vacuolar protein sorting-associated protein 13D; VPS13D |
| Gene Symbols | VPS13D |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.