Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583648 Supplier Invitrogen™ Supplier No. PA583648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AADAFYKAKN EFAAKATEDQ MRLLRLQRRL EDELGGQFLD LSLHDTVTTL ILGGHNKRAE QLARDFRIPD KRLWWLKLTA LADLEDWEEL EKFSKSKKSP IGYLP Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps16 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS16
Gene Accession No. Q9H269
Gene Alias 1810074M16Rik; hVPS16; mVPS16; RP11-12M19.2; vacuolar protein sorting 16 (yeast); vacuolar protein sorting 16 homolog; vacuolar protein sorting 16 homolog (S. cerevisiae); vacuolar protein sorting protein 16; vacuolar protein sorting-associated protein 16 homolog; VPS16; VPS16 CORVET/HOPS core subunit; VPS16, CORVET/HOPS core subunit
Gene Symbols VPS16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS16. Recombinant protein control fragment (Product #RP-99481).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.