Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590792 Supplier Invitrogen™ Supplier No. PA590792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYEPRSGEQV PLLLKMKRSK LALSKAIESG DTDLVFTVLL HLKNELNRGD FFMTLRNQPM ALSLYRQFCK HQELETLKDL YNQDDNHQEL GSFHIRASYA AEERIEGRVA ALQTAADAFY KAKNEFAAKA TEDQMRLLRL QRRLEDELGG QFLDLSLHDT VTTLILGGHN KRAEQLARDF RIPDKRLWWL KLTALADLED WEELEKFSKS KKSPIGYLPF VEICMKQHNK YEAKKYASRV GPEQKVKALL LVGDVAQAAD VAIEHRNEAE LSLVLSHCTG ATDGATADKI QRARAQAQKK; Positive Samples: A-549, Mouse brain, Mouse lung, Rat pancreas; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle, Early endosome, Late endosome membrane, Lysosome membrane, Peripheral membrane protein, autophagosome, clathrin-coated vesicle.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps16 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VPS16
Gene Accession No. Q920Q4, Q9H269
Gene Alias 1810074M16Rik; hVPS16; mVPS16; RP11-12M19.2; vacuolar protein sorting 16 (yeast); vacuolar protein sorting 16 homolog; vacuolar protein sorting 16 homolog (S. cerevisiae); vacuolar protein sorting protein 16; vacuolar protein sorting-associated protein 16 homolog; VPS16; VPS16 CORVET/HOPS core subunit; VPS16, CORVET/HOPS core subunit
Gene Symbols VPS16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 540-839 of human VPS16 (NP_0720972).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296159, 64601, 80743
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.