Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | VPS16 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1785
![]() ![]() |
Proteintech Group Inc
177761AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. In yeast, Vps proteins are involved in the trafficking of endocytic and biosynthetic proteins to the vacuole, which functionally resembles the lysosome of higher organisms. Human VPS16 is the homolog of yeast class C Vps16 protein. VPS16 may play a role in vesicle-mediated protein trafficking to lysosomal compartments and in membrane docking/fusion reactions of late endosomes/lysosomes.Specifications
VPS16 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q920Q4, Q9H269 | |
296159, 64601, 80743 | |
VPS16 Fusion Protein Ag12202 | |
Primary | |
-20°C | |
VPS16 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hVPS16, VPS16 | |
VPS16 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title