Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS18 Antibody (SR1024), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32213750UL
Description
VPS18 Monoclonal antibody specifically detects VPS18 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
VPS18 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1024 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
hVPS18, KIAA1475PEP3, vacuolar protein sorting 18 homolog (S. cerevisiae), vacuolar protein sorting protein 18, vacuolar protein sorting-associated protein 18 homolog | |
A synthesized peptide derived from human VPS18 (Uniprot #: Q9P253) | |
50 μg | |
Autophagy, Cellular Markers | |
57617 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction