Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578546 Supplier Invitrogen™ Supplier No. PA578546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene VPS18
Gene Accession No. Q9P253
Gene Alias 9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit
Gene Symbols VPS18
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VPS18. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.