Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS18 Rabbit anti-Human, Mouse, Rat, Clone: 1W8T9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | VPS18 |
---|---|
Clone | 1W8T9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
VPS18 Monoclonal antibody specifically detects VPS18 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
VPS18 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cellular Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
57617 | |
IgG | |
Affinity purified |
1W8T9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
hVPS18, KIAA1475PEP3, vacuolar protein sorting 18 homolog (S. cerevisiae), vacuolar protein sorting protein 18, vacuolar protein sorting-associated protein 18 homolog | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VPS18 (Q9P253). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title