Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS18 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1953R

Description
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.Specifications
VPS18 | |
Polyclonal | |
Unconjugated | |
VPS18 | |
9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; Vacuolar protein sorting-associated protein 18 homolog; VPS18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit | |
Rabbit | |
Protein A | |
RUO | |
57617 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9P253 | |
VPS18 | |
KLH conjugated synthetic peptide derived from human VPS18. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction