Learn More
Invitrogen™ VPS18 Recombinant Rabbit Monoclonal Antibody (1W8T9)

Description
Positive Samples: HeLa, Mouse lung, Mouse brain, Rat brain Immunogen sequence: MASILDEYEN SLSRSAVLQP GCPSVGIPHS GYVNAQLEKE VPIFTKQRID FTPSERITSL VVSSNQLCMS LGKDTLLRID LGKANEPNHV ELGRKDDAKV.
Specifications
Specifications
| Antigen | VPS18 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1W8T9 |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | VPS18 |
| Gene Accession No. | Q8R307, Q9P253 |
| Gene Alias | 9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.