Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26895425UL
Description
VPS24 Polyclonal antibody specifically detects VPS24 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
VPS24 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
CGI-149, CHMP325.1 protein, Chromatin-modifying protein 3, hVps24, member 3,25.1, NEDFFLJ38114, vacuolar protein sorting 24 (yeast), vacuolar protein sorting 24 homolog (S. cerevisiae) | |
This antibody was developed against a recombinant protein corresponding to amino acids: VNEWSLKIRKEMRVVDRQIRDIQREEEKVKRSVKDAAKKGQKDVCIVLAKEMIRSRKA | |
25 μL | |
Autophagy, Cell Biology, Signal Transduction | |
51652 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction