Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562570
Description
Immunogen sequence: AMSFEWPWQY RFPPFFTLQP NVDTRQKQLA AWCSLVLSFC RLHKQSSMTV MEAQESPLFN NVKLQRKLPV ESIQIVL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Specifications
VPS25 | |
Polyclonal | |
Unconjugated | |
VPS25 | |
1110020N13Rik; AW107467; D11Wsu68e; dermal papilla-derived protein 9; DERP9; Eap20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; HGNC:28122; hVps25; MGC10540; vacuolar protein sorting 25 (yeast); vacuolar protein sorting 25 homolog; vacuolar protein sorting 25 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 25; VPS25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRG1 | |
VPS25 | |
Recombinant protein corresponding to Human VPS25. Recombinant protein control fragment (Product #RP-93263). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction