Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563427
Description
Immunogen sequence: WLDKSKSSFL IMWRRPEEWG KLIYQWVSRS GQNNSVFTLY ELTNGEDTED EEFHGLDEAT LLRALQALQQ EHKAEIITVS D Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Specifications
VPS25 | |
Polyclonal | |
Unconjugated | |
VPS25 | |
1110020N13Rik; AW107467; D11Wsu68e; dermal papilla-derived protein 9; DERP9; Eap20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; HGNC:28122; hVps25; MGC10540; vacuolar protein sorting 25 (yeast); vacuolar protein sorting 25 homolog; vacuolar protein sorting 25 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 25; VPS25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRG1 | |
VPS25 | |
Recombinant protein corresponding to Human VPS25. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction