Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS26A Antibody (CL2287), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$416.50 - $682.00
Specifications
Antigen | VPS26 |
---|---|
Clone | CL2287 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
VPS26A Monoclonal specifically detects VPS26A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
VPS26 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Autophagy | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast) | |
VPS26A | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL2287 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
O75436 | |
9559 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LRKQRTNFHQRFESPESQASAEQPEM | |
Primary | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title