Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS26A Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297752JF549
Description
VPS26A Polyclonal specifically detects VPS26A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VPS26A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast) | |
Produced in rabbits immunized with E. coli-derived Human VPS26A fragment. | |
0.1 mL | |
Autophagy | |
9559 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction