Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS26A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584347 Supplier Invitrogen™ Supplier No. PA584347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRKQRTNFHQ RFESPESQAS AEQPEM Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 96%.

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS26A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vps26a
Gene Accession No. O75436
Gene Alias AA407240; FLJ12930; H beta 58; H>beta>58 protein; HB58; Hbeta58; Hbeta58 homolog; hVPS26; mVPS26; OTTHUMP00000019721; PEP8A; vacuolar protein sorting 26 A; vacuolar protein sorting 26 homolog A; vacuolar protein sorting 26 homolog A (S. pombe); vacuolar protein sorting 26 homolog A (yeast); vacuolar protein sorting-associated protein 26A; vacuole protein sorting 26; vesicle protein sorting 26A; Vps26; VPS26 retromer complex comonent A; VPS26 retromer complex component A; VPS26, retromer complex component A; VPS26A
Gene Symbols Vps26a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS26A. Recombinant protein control fragment (Product #RP-103683).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9559
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.