Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110572 Supplier Invitrogen™ Supplier No. PA5110572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFHGIPATPG IGAPGNKPEL YEEVKLYKNA REREKYDNMA ELFAVVKTMQ ALEKAYIKDC VSPSEYTAAC SRLLVQYKAA FRQVQGSEIS SIDEFCRKFR LDCPLAMERI KEDRPITIKD DKGNLNRCIA DVVSLFITVM DKLRLEIRAM DEIQPDLREL METMHRMSHL PPDFEGRQTV SQWLQTLSGM SASDELDDSQ VRQMLFDLES AYNAFNRFLH A.

This gene encodes a protein involved in endosomal sorting of cell surface receptors via a multivesicular body/late endosome pathway. The encoded protein is one of the three subunits of the ESCRT-I complex (endosomal complexes required for transport) involved in the sorting of ubiquitinated proteins. The two other subunits of ESCRT-I are vesicular protein sorting 23, also known as tumor susceptibility gene 101 (TSG101), and vesicular protein sorting 37. Two alternative transcripts encoding different isoforms have been described. Additional alternative transcripts may exist but the proteins encoded by these transcripts have not been verified experimentally.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS28
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VPS28
Gene Accession No. B5DEN9, Q9D1C8, Q9UK41
Gene Alias 1110014J03Rik; Caspase-activated DNase inhibitor that interacts with ASK1; CIIA; D730005C08Rik; ESCRT-I complex subunit VPS28; H-Vps28; MGC60323; vacuolar protein sorting 28 (yeast); vacuolar protein sorting 28 homolog; vacuolar protein sorting 28 homolog (S. cerevisiae); vacuolar protein sorting 28-like protein; vacuolar protein sorting-associated protein 28 homolog; VPS28; VPS28 subunit of ESCRT-I; VPS28, ESCRT-I subunit; yeast class E protein Vps28p homolog
Gene Symbols VPS28
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human VPS28 (NP_057292.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300052, 51160, 66914
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.