Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS28 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS167925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1679-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1679-25UL Supplier MilliporeSigma Supplier No. ABS167925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-VPS28, Cat. No. ABS1679, is a rabbit polyclonal antibody that detects ESCRT-I complex subunit VPS28 and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS28
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Vacuolar protein sorting-associated protein 28 homolog;H-Vps28;ESCRT-I complex subunit VPS28
Gene Symbols VPS28
Host Species Rabbit
Immunogen A full-length recombinant ESCRT-I complex subunit VPS28 (VPS28).
Molecular Weight of Antigen ∽51 kDa observed; 25.43 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects ESCRT-I complex subunit VPS28 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.