Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518558
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518558 Supplier Invitrogen™ Supplier No. PA518558
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene belongs to a group of vacuolar protein sorting genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS29
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene VPS29
Gene Accession No. Q9UBQ0
Gene Alias 2010015D08Rik; AW049835; DC15; DC7; DKFZp564F0223; EMBL:AAI67055.1, ECO:0000312; FLJ20492; hVPS29; MDS007; PEP11; PEP11 homolog; retromer protein; RGD:1308332}; UniProtKB:Q9UBQ0}; vacuolar protein sorting 29 (S. pombe); vacuolar protein sorting 29 homolog; vacuolar protein sorting 29 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 29; vacuolar protein sorting-associated protein 29 {ECO:0000250; vacuolar sorting protein 29; vacuolar sorting protein VPS29/PEP11; vesicle protein sorting 29; vesicle protein sorting 29 {ECO:0000250; VPS29; vps29 {ECO:0000312; VPS29 retromer complex component; VPS29, retromer complex component; x 007 protein
Gene Symbols VPS29
Host Species Goat
Immunogen Synthetic peptide sequence (CTGNLCTKESYDYLK) corresponding to the internal amino acids of VPS29 (aa 36-51).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51699
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.