Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS33A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531456 Supplier Invitrogen™ Supplier No. PA531456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (83%), Xenopus laevis (86%), Pig (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS33A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Vps33a
Gene Accession No. Q96AX1, Q9D2N9
Gene Alias 3830421M04Rik; AI503300; AW048546; AW554476; bf; Buff; hVPS33A; im:7136893; r-Vps33a; similar to vacuolar protein sorting 33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33 homolog A (S. cerevisiae); vacuolar protein sorting 33A; vacuolar protein sorting 33A (yeast); vacuolar protein sorting protein 33a; vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A; VPS33A core subunit of CORVET and HOPS complexes; VPS33A CORVET/HOPS core subunit; VPS33A, CORVET/HOPS core subunit; wu:fc08g01; wu:fc26c10
Gene Symbols Vps33a
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human VPS33A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65082, 77573
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.