Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS33B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100052
Description
Antibody detects endogenous levels of total VPS33B.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
VPS33B | |
Polyclonal | |
Unconjugated | |
VPS33B | |
FLJ14848; hVPS33B; r-vps33b; vacuolar protein sorting 33 homolog B; vacuolar protein sorting 33 homolog B (yeast); vacuolar protein sorting 33B; vacuolar protein sorting 33B (yeast); vacuolar protein sorting 33-like protein B; vacuolar protein sorting homolog r-vps33b; vacuolar protein sorting-associated protein 33B; vacuolar sorting protein 33b; vps33b; VPS33B late endosome and lysosome associated; VPS33B, late endosome and lysosome associated; zgc:110803 | |
Rabbit | |
Affinity chromatography | |
RUO | |
233405, 26276, 541534, 64060 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59016, Q58EN8, Q63616, Q9H267 | |
VPS33B | |
A synthesized peptide derived from human VPS33B(Accession Q9H267), corresponding to amino acid residues A454-K504. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey, Zebrafish | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction