Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS33B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529139 Supplier Invitrogen™ Supplier No. PA529139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (99%), Rat (98%), Chicken (90%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS33B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene VPS33B
Gene Accession No. P59016, Q9H267
Gene Alias FLJ14848; hVPS33B; r-vps33b; vacuolar protein sorting 33 homolog B; vacuolar protein sorting 33 homolog B (yeast); vacuolar protein sorting 33B; vacuolar protein sorting 33B (yeast); vacuolar protein sorting 33-like protein B; vacuolar protein sorting homolog r-vps33b; vacuolar protein sorting-associated protein 33B; vacuolar sorting protein 33b; vps33b; VPS33B late endosome and lysosome associated; VPS33B, late endosome and lysosome associated; zgc:110803
Gene Symbols VPS33B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VPS33B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233405, 26276
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.