Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS35 Monoclonal Antibody (7E4), PerCP
GREENER_CHOICE

Catalog No. PIMA545938
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545938 Supplier Invitrogen™ Supplier No. MA545938
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45938 was sufficient for detection of VPS35 in 10 μg of SH-SY5Y by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary antibody. Detects approximately 92 kDa.

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS35
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7E4
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene VPS35
Gene Accession No. Q96QK1, Q9EQH3
Gene Alias AI647796; DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; maternal-embryonic 3; Mem3; PARK17; TCCCTA00141; vacuolar protein sorting 35; vacuolar protein sorting 35 homolog; vacuolar protein sorting 35 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35; VPS35 retromer complex component
Gene Symbols VPS35
Host Species Mouse
Immunogen Full length recombinant human VSP35.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25479, 55737, 65114
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.