Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS35 Recombinant Rabbit Monoclonal Antibody (8D3L3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535793
Description
Immunogen sequence: YAGNIIPRLY LLITVGVVYV KSFPQSRKDI LKDLVEMCRG VQHPLRGLFL RNYLLQCTRN ILPDEGEPTD EETTGDISDS MDFVLLNFAE MNKLWVRMQH Q.
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.
Specifications
VPS35 | |
Recombinant Monoclonal | |
0.38 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96QK1, Q9EQH3 | |
VPS35 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-200 of human VPS35 (Q96QK1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
8D3L3 | |
Unconjugated | |
VPS35 | |
AI647796; DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; maternal-embryonic 3; Mem3; PARK17; TCCCTA00141; vacuolar protein sorting 35; vacuolar protein sorting 35 homolog; vacuolar protein sorting 35 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35; VPS35 retromer complex component | |
Rabbit | |
Affinity chromatography | |
RUO | |
25479, 55737, 65114 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction